Would you like to log in again?
PHCbi MPR Series Pharmaceutical Refrigerators, PHC Corporation
Supplier: PHC Corporation
PHCbi
MPR-S300H-PA MPR-S300H-PA MPR-S500H-PA MPR-S500RH-PA MPR1014PA MPR1014RPA MPR1014RPA MPR1014PA MPR-S500H-PA MPR-S500RH-PA
76518-110EA 8664.96 USD InStock
CA76383-298 CA76518-112 CA97035-150 76518-110 76383-298 CA76518-110 76518-112 CA97035-152 97035-152 97035-150
PHCbi MPR Series Pharmaceutical Refrigerators, PHC Corporation
Refrigerators
PHCbi brand Pharmaceutical Refrigerators offer a complete and integrated solution for the increased requirements of achieving strict and regulated storage temperatures for pharmaceuticals, medicines, vaccines, and other temperature-sensitive biological samples.
  • Integrated solution
  • Slim design
  • High-efficiency compressor
  • 2 year manufacturer's parts and labor warranty
  • 3 year manufacturer's compressor parts warranty
The slim front-to-back design and optional sliding shelves allow for ergonomic and accessible product retrieval, while fitting seamlessly into any facility.
Units utilize a highly efficient hermetic compressor to provide efficient cooling and maintain set temperature levels. The thermistor sensor, microprocessor, and electronic temperature controls function to ensure that the set temperature is maintained. Even with frequent door openings, the circulation fan provides rapid temperature adjustment for a highly reliable and stable preservation environment unaffected by ambient temperature. Rigid polyurethane foam insulation construction acts to keep cold air inside the unit and effectively protects against the intrusion of warm air.
Defrosting is performed automatically during compressor “off” cycles by sensing frost levels. This way defrosting is performed only when required, further protecting the contents against unnecessary temperature rises. The defrosting heater also acts as an emergency heat source to prevent samples from freezing in extremely low ambient temperature conditions (below 0 °C).
Ordering information: All units are supplied with a 2 year manufacturer's parts and labor warranty and a 3 year manufacturer's compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty