Would you like to log in again?
AcroPak™ 20 Filtration Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AcroPak™
12202 12203 12247 12200 12247 12201 12200 12202 12203 12246 12246 12201
28143-841PK 68.62 USD InStock
28143-841 28143-843 97035-168 CA28143-841 28143-845 CA28143-843 97035-166 28143-847 CA28143-845 CA97035-166 CA28143-847 CA97035-168
AcroPak™ 20 Filtration Capsules, Cytiva (Formerly Pall Lab)
Filters Capsule Filters
AcroPak™ 20 capsule filters are designed to efficiently process up to 2 litres of chemical and biological fluids. These filters are available in three different membrane types.
Supor®
The Supor® membrane has high flow rates and throughputs, and is ideal for solutions where low protein binding is required. Not recommended for use with some ketones. For sterile filtration of media and buffers.
• Built-in prefilter extends filter life when particulate-laden solutions such as serum-containing media are processed
• Sterilisation by gamma irradiation eliminates potential contamination by EtO residuals
• PP housing and vent plug, PC filling bell, Supor® (hydrophilic PES) membrane
• Suitable for use with fluids containing dilute proteins, preservatives, or other critical components
Supor® EKV
Built-in MachV asymmetric pre-filter layer for maximum flow and throughput performance. For Sterile filtration of media and buffers.
• Disposable for reduced labour and associated costs
• Significantly reduces the cost of repeat testing when scaling up your filtration system
• Ideal for filtering aqueous solutions, cell culture media and serum
• Suitable for use with fluids containing dilute proteins, preservatives, or other critical components
Fluorodyne® II
The Fluorodyne® II membrane is compatible with aqueous and many organic solvents.
• Double layer sterilising membrane enhances reliability
• Upstream air vent prevents vapour lock
• Manufactured without the use of adhesives to minimise extractables
Supor and Supor EKV can withstand one autoclave cycle at 125 ºC (257 ºF) for 60 min; water wet capsule prior to autoclaving. Fluorodyne II can withstand one autoclave cycle at 131 °C (268 °F) for 30 min.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty