Would you like to log in again?
VWR® Thermometer Armor Cases
Supplier: VWR
VWR®
61400-9511 61400-9311 20930 61400-9611
61222-686EA 25.56 USD InStock
61222-686 97035-494 82007-988 61222-702
VWR® Thermometer Armor Cases
Thermometer Holders
These nickel-plated brass armor cases protect thermometers from accidental breakage during use and storage.
Lower section surrounding bulb has small openings for circulation
Available in five lengths to fit SAMA general-purpose total and partial immersion thermometers, including fluoropolymer resin-coated thermometers.
Fitted with screw cap and ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty