Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Silicone Safety Pipette Filler, Sper Scientific
Supplier: Sper Scientific
470114 470113
97035-726EA 45.29 USD InStock
97035-726 97035-724
Silicone Safety Pipette Filler, Sper Scientific
Pipettes Pipette Bulbs
Pipette fillers enable filling and dispensing of pipettes with one hand.
  • Adjusts to fit any size pipet
  • Chemically resistant glass valves
  • Available in two colors for identification purposes
Safety is ensured through avoidance of hand or mouth contact with liquid. Our Silicone Pipette Fillers offer a softer feel with easier and faster operation of valves. Silicone is also more chemically resistant and more resilient than either PVC or natural rubber. It will not crack or become brittle and will last longer and stay flexible.
Ordering information: Glass tube not included, order separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty