Would you like to log in again?
Water Resistant Stopwatch, Sper Scientific
Supplier: Sper Scientific
810012C 810012
97035-842EA 116.62 USD InStock
97035-842 97035-840
Water Resistant Stopwatch, Sper Scientific
Timers Digital Stopwatches
Water resistant digital stopwatch.
  • Split interval function
  • LCD display
Counts up to 24 hours in ¹/₁₀₀th second resolution for the first 30 minutes with a 1 second resolution thereafter. Also has a 12/24 hour alarm clock, calendar and an hourly chime option.
Certifications: N.I.S.T. traceable certificate of compliance for 97035-842.
Ordering information: Five year warranty.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty