Would you like to log in again?
Observational Research Stopwatch, Sper Scientific
Supplier: Sper Scientific
810029AR 810029ARC
97035-866EA 122.07 USD InStock
97035-866 97035-864
Observational Research Stopwatch, Sper Scientific
Timers Digital Stopwatches
A unique timer which allows the timing of a subject without disturbing it.
  • For captive and field research, or in classrooms
  • Hands free operation allows for freedom of movement
  • Counts up or down to 10 hours
  • Shock resistant, with good hand grip
  • Ideal for multiple and simultaneous processes
The alarm is audible only through a headphone. The alarm can be set for repetitive intervals of every 0.25 second to every hour. Two ¹/₁₀₀th second displays. In stopwatch mode current lap time shown in upper display, cumulative total in lower one. Memory stores last 8 lap times so there is no need to look away from a test to make notes. In timer mode upper display shows original setting while lower one counts down. Warning alarm sounds at 30 seconds before time elapses and when time has elapsed.
Countdown cycles repeat upon completion. 30 second shuttle timer for use during timeouts. Lap counter shows number of laps (up to 99) in stopwatch mode and number of cycles repeated in timer mode. Pacesetter function can be set to beep continually from every 0.25 second to every 30 seconds.
Certifications: N.I.S.T. traceable certificate of compliance 97035-866.
Ordering information: Five year warranty.
Delivery information: Includes earpiece and battery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty