Would you like to log in again?
Continuous Alarm Timer, Sper Scientific
Supplier: Sper Scientific
810045C 810045
97035-906EA 135.3 USD InStock
97035-906 97035-904
Continuous Alarm Timer, Sper Scientific
Timers Digital Timers
Select either a bright flashing yellow LED visual alarm or an audible alarm.
A memory recalls the last timer setting for reuse. Built in stand for bench top use, clips to a pocket, or sticks to any metal surface with a magnetic back.
Certifications: N.I.S.T. traceable certificate of compliance for 97035-906.
Ordering information: Five year warranty.
Delivery information: Two AAA batteries included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty