Would you like to log in again?
Large Display Bench Timer, Sper Scientific
Supplier: Sper Scientific
810046 810046C
97035-910EA 111.89 USD InStock
97035-910 97035-908
Large Display Bench Timer, Sper Scientific
Timers Digital Timers
Although small and easily portable, this timer features a large display with ³/₄" high digits.
  • Counts down from 100 minutes with 1-second resolution
  • Has a built in stand, pocket clip and magnetic back
Also unique is an auto-power-off feature to extend battery life (operates only when the timer is idle for 10 minutes and will not interrupt a timing sequence). Short warning alarms sound at 3 and 5 minutes prior to the final 30-second alarm. Automatically records elapsed time following the alarm for up for to 20 minutes, or until stopped. A memory recalls the last timer setting for repetitive tests.
Certifications: N.I.S.T. traceable certificate of compliance for 97035-910.
Ordering information: Five year warranty.
Delivery information: One AAA battery included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty