Would you like to log in again?
Vacuum Meters, Sper Scientific
Supplier: Sper Scientific
840064 840064C
97035-950EA 926.88 USD InStock
97035-950 97035-948
Vacuum Meters, Sper Scientific
Pumps Vacuum Gauges
Portable meter for absolute vacuum pressure measurements in hospital labs, crystal growing, solvent recovery, distillation, and rotary evaporation
Also used in HVAC, automotive and other industrial applications. Includes a detachable heavy-duty probe with ¼ PS connector for liquid, air, oil and gas. Features a large display with eight units of measure, min/max, hold, low battery indicator, auto power off and an RS232 computer interface.
Certifications: N.I.S.T. traceable certificate of compliance (for certified model only).
Ordering information: 1-year warranty. Comes ready to use in a foam-lined, hard-shell carrying case, with 9V battery and instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty