Would you like to log in again?
Advanced pH Meter, Sper Scientific
Supplier: Sper Scientific
850055
97036-026EA 467.79 USD InStock
97036-026
Advanced pH Meter, Sper Scientific
Electrochemical Meters pH and Multi-Parameter Meters
Benchtop level analysis in the field.
Reads either pH (in 0.01 resolution) or mV in large digits, with time, date, and temperature (in °C or °F) shown simultaneously on the lower display. Also indicates stable reading, low battery and displays calibration data. Scroll 99 data points with time stamps and min/max readings directly on the backlit LCD. Features hold, up to 5 point calibration, automatic buffer recognition, manual or automatic temperature compensation, and a USB port. Will accept any pH or ORP probe with a BNC connector.
Ordering information: Probe not included. Comes with 4 AAA batteries.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty