Would you like to log in again?
HIGHPlate* 6000 Microplate Rotor, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75003606 75003606
97040-244EA 9353.3 USD InStock
CA97040-244 97040-244
HIGHPlate* 6000 Microplate Rotor, Thermo Scientific
Centrifuges Rotors
Thermo Scientific* HIGHPlate 6000 Microplate Rotor spins filter plates.
The rotor accommodates a variety of filtration plates up to 80mm high. 2x8 deepwell plates.
Recommended for: DNA purification protocols.
Certifications: Biocontainment certified by CAMR*, HPA, Porton Down, U.K.
Ordering information: Compatible with Thermo Scientific Nalgene bottles and tubes for complete centrifuge solutions. Compatible for Thermo Scientific Multifuge X1/X3/X3 F Series.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty