Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Highconic™ II Aluminum Fixed-Angle Rotor, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75003620 75003620
97040-260EA 5514.28 USD InStock
CA97040-260 97040-260
Highconic™ II Aluminum Fixed-Angle Rotor, Thermo Scientific
Centrifuges Rotors
Thermo Scientific* HIGHConic II Aluminum Fixed-Angle Rotor delivers biocontainment at 10,350rpm.

Capacity flexibility with adapters ranging from 564 to 48mL. Choose from round or conical tubes with a wide array of available adapters.

Certifications: Biocontainment certified by CAMR*, HPA, Porton Down, U.K.

Ordering information: Adapters are sold separately. Replacement sealing O-rings are also available (in sets of 2 with grease). Contact your Sales Representative for more information. Compatible with Thermo Scientific Megafuge 40/40R, Multifuge X3/X3R X3 F/X3 F R Centrifuge Series.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty