Would you like to log in again?
Fiberlite™ F13-14x50cy Fixed-Angle Rotor, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
75003661 75003661 46922
97040-272EA 7119.38 USD InStock
97040-272 101623-000 CA97040-272
Fiberlite™ F13-14x50cy Fixed-Angle Rotor, Thermo Scientific
Centrifuges Rotors
The Fiberlite™ F13-14x50cy Fixed-Angle Rotor provides 14-place capacity and high maximum RCF.
These lightweight, carbon fiber rotors maximize performance with versatility, speed and a robust, corrosion-free design.
Enhanced productivity: Reduce run-times with higher G-forces with our lightweight carbon fiber rotors.
Unequalled durability: Corrosion-and fatigue-resistant design ensures structural integrity.
Secure investment: Backed by a 15-year warranty.
Recommended for Pelleting prokaryotic and eukaryotic cells from cell cultures.
Certifications: Biocontainment certified by CAMR*, HPA, Porton Down, U.K.
Ordering information: Tube adapters are sold separately. For O-ring refresher kit contact your Sales Representative for more information. Compatible with Thermo Scientific Multifuge X3/X3R/X3F/X3FR Centrifuge Series.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty