Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Fiberlite™ F14-6x250LE Fixed-Angle Rotor, Thermo Scientific
Supplier: Thermo Scientific
75003662 75003662
97040-274EA 9146.43 USD InStock
CA97040-274 97040-274
Fiberlite™ F14-6x250LE Fixed-Angle Rotor, Thermo Scientific
Centrifuges Rotors
The Fiberlite™ F14-6x250LE Fixed-Angle Rotor supports a range of capacities and applications
  • Reduced run-times with higher G-forces achieved through lightweight carbon fiber rotors
  • Corrosion- and fatigue-resistant design that ensures structural integrity
  • Compatible with Thermo Scientific™ Nalgene™ bottles and tubes
  • Backed by a 15-year warranty
Recommended for pelleting bacteria or yeast cells and for purifying nucleic acids and cells.
The lightweight, carbon fiber rotor maximizes performance with versatility, speed and a robust, corrosion-free design.
Certifications: Biocontainment certified by CAMR, HPA, Porton Down, U.K.
Ordering information: Tube adapters are sold separately. For O-ring refresher kit contact your Sales Representative for more information. Compatible with Thermo Scientific Multifuge X3/X3R/X3F/X3FR Centrifuge Series.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty