Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AcroPak™ Filtration Capsules, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

AcroPak™
12461 12999 12686 12471 12467 12995 12460 12094 12993 12471 12461 12675 12464 12473 12994 12991 12478 12686 12466 12999 12093 12095 12069 12994 12094 12997 12069 12095 12995 12996 12093 12996 12675 12466 12992 12473 12464 12993 12941 12460 12997 12467 12463 12992 12463 12478 12941 12991
97040-968EA 212.8 USD InStock
97040-968 28143-820 CA97035-170 28143-864 97040-966 CA97035-172 97040-964 87003-890 28143-824 97040-962 28146-178 CA97040-972 28146-179 28143-969 28146-198 CA28146-185 28146-177 28143-828 CA97040-970 CA28146-180 28146-196 CA28146-183 CA28146-182 CA28146-181 CA97040-968 CA97040-966 CA97040-964 CA28143-820 CA28143-864 CA97040-962 CA28143-824 CA28146-177 CA28146-198 CA87003-890 CA28143-969 CA28146-196 97040-972 CA28143-828 97040-970 CA28146-179 CA28146-178 28146-181 28146-182 28146-180 28146-185 28146-183 97035-172 97035-170
AcroPak™ Filtration Capsules, Cytiva (Formerly Pall Lab)
Filters Capsule Filters
A filter for every step in your process. From drug discovery and basic research to process development and production, Pall Laboratory’s AcroPak capsules come in a variety of membrane types and volume sizes.

  • 100% integrity tested (where noted)
  • Endotoxin level: <0.25 EU/ml using Limulus Amebocyte Lysate test
  • Bacterial retention: 0.2 μm lot samples retain 10⁷ cfu/cm² of B. diminuta per modified ASTM F838
  • Mycoplasma reduction: 0.1 μm lot samples retain 10⁷ cfu/cm² A. Laidlawii per modified ASTM F838
  • Biological safety: Passes United States Pharmacopeia (USP) biological reactivity test in vivo <88>
  • Sterilisation by gamma irradiation

Primary applications include: Buffer media and reagent manufacturing, protein purification and cell culture and bioburden filtration.

Supor PES (Polyethersulfone):
With high flow and capacity, broad fluid compatibility, low protein adsorption, active ingredients, and stabilizers, Pall PES membranes are best suited for filtration of preparative fluids such as buffers, media, and bioprocess intermediates.

Supor EKV Asymmetric PES:
PES filters with asymmetric membrane layers have the same benefits of Supor PES (above), but with additional debris holding. This makes them ideal for high turbidity, high bioburden solutions. All Supor EKV products are a 0.65 µm pre-filter and a 0.2 µm asymmetric membrane.

Fluorodyne II PVDF (hydrophilic polyvinylidene fluoride):
Low extractables, low product and excipient adsorption, simple validation for the sterilization of a wide range of formulations make PVDF membranes ideal for final filtration.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty