Would you like to log in again?
VWR® Basic SPP Shoe Covers
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
40GSM XL 414004-653 40GSM U 97041-234 97041-236
414004-653CS 114.1 USD InStock
414004-653 89233-796 97041-234 89233-794 97041-236
VWR® Basic SPP Shoe Covers
Disposable Garments
These spunbonded polypropylene (SPP) shoe covers protect against dirt, grime, and certain dry particulates in nonhazardous environments
  • Economical, disposable first-line barrier
  • Maximum breathability
  • Low linting
Available with rubber-coated soles for secure footing on wet or dry surfaces. Elastic top and bottom. Shoe covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
The fabric is strong, lightweight, and breathable. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty