Would you like to log in again?
ScienceLine pH Combination Electrodes with Temperature Sensor, SI Analytics
Supplier: SI Analytics
285130200 285105476 285130240 285093111 1054512 285100380 285100510 285130210 285100531 285093009 285130280 285100500 285130260 285130270 285130250
97042-128EA 1200.34 USD InStock
97042-128 97042-126 97042-138 97042-076 97042-142 97042-074 97042-140 97042-350 97042-136 97042-430 97042-078 97042-134 97042-080 97042-072 97042-062
ScienceLine pH Combination Electrodes with Temperature Sensor, SI Analytics
Electrodes pH Electrodes
These pH combination electrode have MMR tubes with a diameter of 3 mm and a length of 180 mm
They feature round bottom flask with NS 14.5 and NS 7.5 standard taper.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty