Would you like to log in again?
Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Supplier: SI Analytics
285138538 285138295
97042-510EA 202.81 USD InStock
97042-510 97042-452
Cleaning Solution, Pepsin/Hydrochloric Acid Solution, SI Analytics
Cleaning Agents Detergents and Surface Cleansers
Suitable for cleaning combination electrodes and reference electrodes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty