Would you like to log in again?
SenTix® pH Electrodes, WTW
Supplier: WTW
103652 103647 103641 103663 103655 103633 103639 103700 103651 103642 103637 103695 103640 103696 103645 103635 103667 103636 103643 103661 103631 103702 103669 103690 103630 103646 103660 103650 103668
97042-830EA 1641.39 USD InStock
97042-830 97042-798 97042-814 97042-792 80085-228 97042-796 97042-794 80085-234 80085-212 80085-232 80085-210 80085-216 80085-214 97042-806 80085-230 97042-822 97042-800 97042-820 97042-826 97042-824 97042-802 80085-218 80085-222 80085-220 80085-226 80085-224 97042-818 97042-816 80085-240
SenTix® pH Electrodes, WTW
Electrodes pH Electrodes
Low-resistance glass membranes guarantee stable measuring signals even at low temperatures
Low-maintenance pH electrodes with gel electrolyte are ideal for portable measurements, as well as for routine measurements in-the-laboratory. With or without built-in temperature probe all electrodes have robust plastic shafts with a low-maintenance gel reference system.
Special electrodes for unique applications need special electrodes. SenTix special electrodes can take on the challenges associated with measuring the pH value of surfaces, solids, suspensions, emulsions, low ionic samples, smallest volumes and more. For those who require a non-glass electrode, the SenTix FET can be used with every WTW pH meter.
Silver ion-free reference electrolyte, together with the proven platinum wire diaphragm, prevent measurement problems b precipitating silber compounds. Functional slide for accessing te refill opening for electrodes with liquid electrolyte.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty