Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
pH Electrodes, IDS (Intelligent Digital Sensors), for Multi, WTW
Supplier: WTW
IDS®
103750 103740 103741 103780
97042-844EA 825.02 USD InStock
97042-844 97042-836 97042-846 97042-838
pH Electrodes, IDS (Intelligent Digital Sensors), for Multi, WTW
Electrodes pH Electrodes
High quality, enhanced sensor technology combined with most modern measurement electronics.
  • IDS electrodes include their serial number and their calibration records
  • Measurement integrity benchmarking of the cells by QSC (Quality Sensor Control)
  • Ø×L: 12×120 mm
Ordering information: Various cable lengths available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty