Would you like to log in again?
Combined Ion Selective Electrodes, 800 Series, WTW
Supplier: WTW
106660 106674 106664 106663 106667 106666 106662 106661 106658 106650 106654 106665 106670 106668
97042-888EA 4602.59 USD InStock
97042-888 97042-886 80086-698 97042-884 97042-882 80085-300 80085-298 80085-286 80085-304 80085-292 80085-290 80085-296 80085-294 80085-282
Combined Ion Selective Electrodes, 800 Series, WTW
Electrodes ISE Electrodes
These easy-to-use, robust, high quality ion-selective electrodes are ideal for field applications.
Combined with built-in reference electrodes which make them particularly suitable for measurement of small volume samples.
Ordering information: These ion selective electrodes will require additional electrolyte and ion strength adjustment solutions (ISA) as well as freshly prepared standards (see ISA solutions).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty