Would you like to log in again?
Conductivity Cells, IDS, for Multi 3410 / 3420 / 3430, WTW
Supplier: WTW
301711 301710 301720
97042-954EA 1375.76 USD InStock
97042-954 97042-952 97042-960
Conductivity Cells, IDS, for Multi 3410 / 3420 / 3430, WTW
Electrodes Conductivity Sensors
High quality, enhanced sensor technology combined with most modern measurement electronics.
  • IDS cells include their serial number and their calibration records
  • Two different cells covering all applications from pure water to highly conducting solutions
  • Temperature Range: 0 to +100 °C
Various cable lengths available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty