Would you like to log in again?
ProfiLine™ Single-Parameter Portable Meters, pH/mV, WTW
Supplier: WTW
ProfiLine™
2AA110
97043-094EA 1366.27 USD InStock
97043-094
ProfiLine™ Single-Parameter Portable Meters, pH/mV, WTW
Electrochemical Meters pH and Multi-Parameter Meters
Meters provide reliable measurements for field applications
The robust Model pH 3110 meter provides easy operation in the field. The versatile Model pH 3210 meter includes backlit graphic LCD, automatic stability control (AutoRead), an internal clock, memory that stores up to 200 data sets, and Continuous Measurement Control (CMC) function that ensures measurement within a specified calibration range.
Equipped with built-in calibration indicator and waterproof to IP67 standards.
Ordering information: Supplied with short instruction manual, CD-ROM, and batteries. Meter kit 97043-104 includes a SenTix® 41 pH probe, buffer solutions STP 4 and STP 7, carry case, stand, and beaker.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty