Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Standard Heavy-Duty Vortex Mixer
Supplier: VWR
VWR®
80862961
97043-562EA 1367.81 USD InStock
97043-562
VWR® Standard Heavy-Duty Vortex Mixer
Shakers and Mixers Vortex Mixers
Variable speed analog mixer features a heavy-duty design and efficient motor that permits continuous duty operation and the ability to handle accessory components over the entire speed range.
  • Designed for continuous duty
  • Now with Best-in-Class, Exclusive 5-Year Warranty
Users can select from two distinct operating modes; "On" mode enables use with any of the accessory attachments during continous operation. Alternately, "Touch" mode is designed to facilitate use with the cup head or universal holder with cover.
Certifications: RoHS certified. CSA certified. The 230V units are CE marked.
Ordering information: Supplied with a cup head, universal holder with cover, foam insert for thirty-eight 1.5 to 2.0 mL microtubes, and a 234 cm (92") detachable, 3-wire cord and plug. 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty