Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Advanced Heavy-Duty Vortex Mixer
Supplier: VWR
VWR®
80862959 80862960
10027-194EA 1689.62 USD InStock
10027-194 97043-564
VWR® Advanced Heavy-Duty Vortex Mixer
Shakers and Mixers Vortex Mixers
Ideal for applications that require repeatable results, the mixer features touchpad controls and LED displays for accurate speed and time results.
  • Designed for continuous duty
  • LED displays for time and speed
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
A heavy-duty design and efficient motor permit continuous duty operation and the ability to handle accessory components over the entire speed range. Users can select from two distinct operating modes; "On" mode enables use with any of the accessory attachments during continous operation. Alternately, "Touch" mode is designed to facilitate use with the cup head or universal holder with cover. Microprocessor control maintains the set speed for strong, consistent mixing action. The timer will display elapsed time or, when set to a defined limit, will shut off the unit when time reaches zero.
Certifications: RoHS certified. CSA listed. Calibrated Vortex Mixers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple.
Ordering information: Supplied with a cup head, universal holder with cover, foam insert for thirty-eight 1.5 to 2.0 mL microtubes, and a 234 cm (92") detachable, 3-wire cord and plug (230V units are supplied with a Euro-type plug). 230V, 50/60Hz models are also available; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty