Would you like to log in again?
Protective Covers for NewClassic MS Series High Capacity Balances, METTLER TOLEDO®
Supplier: Mettler-Toledo
NewClassic
12121851 12121852 12121851 12121850 12121853 12121853 12121852 12121850
97044-178EA 75.92 USD InStock
97044-178 CA97044-176 97044-176 CA97044-182 97044-182 CA97044-180 97044-180 CA97044-178
Protective Covers for NewClassic MS Series High Capacity Balances, METTLER TOLEDO®
Balances and Scales
These protective covers decreases the need to clean your balance and helps prevents cross contamination.
  • Protects the balance against toxic or hazardous substances and provides operators with an increased level of safety
  • The cover is installed and removed in a few simple steps. Once in place, it does not interfere with balance operation
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty