Would you like to log in again?
BELOW BALANCE HOOK FOR MS-L
Supplier: Mettler Toledo
Mettler Toledo Hook^ For Newclassic Ms-L Balances, Up To 1G. Enables Weighing Below The Balance.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BELOW BALANCE HOOK FOR MS-L 97044-184 11132565 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty