Would you like to log in again?
DOOR DRAFT SHIELD WITH SLOT FOR XPE/XS
Supplier: Mettler Toledo
Door Draft Shield With Slot For Xpe/Xs^ Draft shield door, with slot for the LV11 feeder for XPE/XS analytical balances, Faster weighing, Easy Installation, Protecting weighing samples from surrounding influences by minimizing air turbulence ensures faster stabilization
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
DOOR DRAFT SHIELD WITH SLOT FOR XPE/XS 97044-252 11106715 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty