Would you like to log in again?
Economical Stainless Steel Slotted Weight Sets
Supplier: Troemner
30390750 30390731 30390729 30390745 30390735 30390733
10754-320EA 35 USD InStock
10754-320 97044-288 97044-286 97044-284 97044-292 10754-288
Economical Stainless Steel Slotted Weight Sets
Weights Calibration Weights
The makeup of economical stainless steel weight sets varies.
Slotted weights 10 to 500 g (0.1 to 5 N) are made of stainless steel, and weights 1 to 5 g are made of aluminum. All slotted weights have an approximate diameter of 1⁵/₈" and have a slot opening width of ¹/₄". A hanger is also available from which you can stack and hang your slotted weights. The overall length of the hanger is approximately 7⁷/₈" with a usable length of 7". The hanger is designed to accept the slotted weights.
Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty