Would you like to log in again?
Economical Individual Weights, Troemner
Supplier: Troemner
30391017 30391014 30391001 30391004 30391010 30391007 30391009 30391018 30391003 30390999 30391000 30391013 30391002 30391015 30391012 30391008 30391016 30391005 30391006 30391011
97044-320EA 28.14 USD InStock
97044-320 97044-298 97044-310 97044-296 97044-294 97044-306 97044-318 97044-326 10754-348 97044-304 97044-316 97044-324 97044-302 97044-314 97044-322 97044-300 97044-312 10754-346 10754-344 97044-308
Economical Individual Weights, Troemner
Weights Calibration Weights
Individual weights are available from 2 kg to 1 mg.
Weights 1 g and above are carefully manufactured of stainless steel and are cylindrical in shape for easy gripping. Weights 1 g to 100 g are made from one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight.
Weights 200 g through 2 kg have an adjusting cavity on the top that is sealed with a plug. Economical stainless steel weights 1 g and above are perfectly flat on top to stack easily on center.
Weights 500 mg and lower are made of sheet metal. They feature one-piece construction with one side turned up to make them easy to handle with forceps. The 500 mg and 200 mg weights are made of stainless steel while weights 100 mg and below are made of aluminum.
Certifications: Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty