Would you like to log in again?
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Covers, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
07-6060N 07-2065N 07-8415N 07-3010N 07-8061N 07-2011N 07-3066N 07-1265N 07-1266N 07-8461N 07-3063N 07-7060N 07-1063N 07-1111N 07-1263N 07-2066N 07-1010N 07-6061N 07-5015N 07-8460N 07-5060N 07-1163N 07-8015N 07-6067N 07-8067N 07-3911N 07-7015N 07-3910N 07-8060N 07-2063N 07-8017N 07-1066N 07-3963N 07-1211N 07-2010N 07-1166N 07-3011N 07-3065N 07-1011N 07-7417N 07-7467N 07-7460N 07-7461N 07-7415N 07-6015N 07-1210N 07-5017N 07-8417N 07-1110N 07-1165N 07-7067N 07-5061N 07-6017N 07-7017N 07-7061N 07-5067N 07-1065N 07-3965N 07-8467N
97044-942EA 33.7 USD InStock
97044-942 97044-940 97045-350 97045-274 97045-276 97045-192 97044-938 97045-194 97045-030 97045-272 97045-358 97045-876 97045-877 97045-878 97045-872 97045-356 97045-874 10806-034 97044-934 97044-932 97045-361 97045-362 97044-928 97044-926 97044-924 97045-360 97045-326 97045-328 97045-324 97045-329 97044-920 97045-330 97044-918 10806-072 97045-336 97045-338 97045-339 10806-070 97045-334 10806-054 97044-950 97045-064 97045-340 97045-142 97045-144 97044-948 97045-380 97045-062 97045-382 97045-028 97045-348 97045-744 97045-745 97045-349 97045-746 97045-146 97045-344 97045-026 97045-346
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Covers, DWK Life Sciences
Microplates Deep Well Microplates
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with covers.
  • Variety of volumes available
  • EVA covers offer superior seal
  • Silicone mats are resealable
The 96-well round and square well microplates are available in a variety of volumes. Constructed of natural polypropylene, all microplates are made to ANSI specifications for 96-well microplates. Covers made of Ethylene Vinyl Acetate (EVA) are piercible by standard needle configurations found on most HPLC, LC/MS, CombiChem, and HTS instruments. They have a superior seal compared to similar products. For users who require some level of resealability, two silicone mats with a sprayed-on fluoropolymer resin barrier are available. Silicone mats are available in standard and pre-cut formats.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty