Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and Caps, Assembled, DWK Life Sciences
Supplier: DWK Life Sciences
07-8075N-1000-A 07-8045N-2200-A 07-8075R-1202-A 07-8020R-1202-A 07-3045G-350-A 07-2045B-201-A 07-8021G-1200-A 07-8070N-1000-A 07-1220R-101-A 07-2021R-201-A 07-3070G-350-A 07-3021N-300-A 07-8070R-1000-A 07-8075N-2200-A 07-8075G-1200-A 07-8045B-1000-A 07-8021B-1000-A 07-2021G-201-A 07-8020G-1000-A 07-1221B-101-A 07-3020B-350-A 07-3045B-300-A 07-8075G-1202-A 07-8020R-1000-A 07-8020R-1200-A 07-8070R-1202-A 07-8075N-1202-A 07-8021G-2200-A 07-3021G-300-A 07-8075N-1200-A 07-1221N-101-A 07-8075R-2200-A 07-2020R-250-A 07-2021R-250-A 07-3045R-350-A 07-2021N-250-A 07-8020B-2200-A 07-8021N-1202-A 07-3075N-300-A 07-3020N-350-A 07-2020B-250-A 07-8075R-1000-A 07-8045B-2200-A 07-8070N-1200-A 07-2070R-201-A 07-8070G-1200-A 07-8020B-1200-A 07-8075B-1000-A 07-3020B-300-A 07-3021B-350-A 07-3045N-350-A 07-2070G-250-A 07-2021B-250-A 07-2021N-201-A 07-8020G-2200-A 07-2075G-250-A 07-8075G-1000-A 07-8070B-1200-A 07-2075R-250-A 07-2070B-250-A 07-2075B-201-A 07-3075N-350-A 07-8020G-1202-A 07-8075B-2200-A 07-2075R-201-A 07-1275B-101-A 07-3045R-300-A 07-8020B-1202-A 07-8045N-1000-A 07-2070N-201-A 07-8070B-1000-A 07-3021N-350-A 07-1270R-101-A 07-2075G-201-A 07-8045R-1000-A 07-3020R-300-A 07-3070B-350-A 07-2075N-201-A 07-2020G-201-A 07-2045R-250-A 07-8075B-1200-A 07-1245R-101-A 07-2045N-201-A 07-2020G-250-A 07-3020N-300-A 07-8075R-1200-A 07-2075B-250-A 07-8045R-2200-A 07-8045N-1202-A 07-2021B-201-A 07-3045N-300-A 07-3070R-300-A 07-3020G-350-A 07-1275G-101-A 07-8045B-1202-A 07-8020B-1000-A 07-8070N-1202-A 07-8021R-1000-A 07-3021R-300-A 07-3045B-350-A 07-8070N-2200-A 07-2070G-201-A 07-8020G-1200-A 07-8021B-1202-A 07-2070N-250-A 07-8020N-1000-A 07-3075B-300-A 07-3075G-300-A 07-8020R-2200-A 07-1220MB-101 07-3075R-350-A 07-8070B-2200-A 07-1270N-101-A 07-8020N-2200-A 07-8021N-1000-A 07-2021G-250-A 07-8021G-1000-A 07-3075G-350-A 07-8045G-1000-A 07-2075N-250-A 07-3070B-300-A 07-8045G-1202-A 07-1245G-101-A 07-1221R-101-A 07-1220G-101-A 07-3021G-350-A 07-8045R-1202-A 07-8070R-2200-A 07-8021N-2200-A 07-1220N-101-A 07-2045G-201-A 07-2045B-250-A 07-8021G-1202-A 07-2070R-250-A 07-3020R-350-A 07-8045R-1200-A 07-8070G-1000-A 07-2070B-201-A 07-8070G-1202-A 07-2020B-201-A 07-3021R-350-A 07-1221G-101-A 07-8045B-1200-A 07-3070N-300-A 07-3070G-300-A 07-3075B-350-A 07-8045N-1200-A 07-2020N-250-A 07-3070N-350-A 07-1275N-101-A 07-3075R-300-A 07-8075G-2200-A 07-8020N-1200-A 07-2045G-250-A 07-8021B-2200-A 07-8021N-1200-A 07-8045G-2200-A 07-2020R-201-A 07-3021B-300-A 07-8075B-1202-A 07-8021R-2200-A 07-8070B-1202-A 07-1245N-101-A 07-8021R-1202-A 07-8020N-1202-A 07-1275R-101-A 07-3020G-300-A 07-8070R-1200-A 07-1270G-101-A 07-3045G-300-A 07-8021B-1200-A 07-2020N-201-A 07-2045R-201-A 07-3070R-350-A 07-8070G-2200-A 07-1270B-101-A 07-2045N-250-A 07-8021R-1200-A 07-8045G-1200-A 07-1245B-101-A
97044-980PK 162.05 USD InStock
97044-980 89231-806 97045-706 97044-984 97045-394 97045-154 97045-034 30618-350 97045-270 30618-232 97045-150 30618-354 89231-930 30618-236 30618-358 97045-398 97045-830 97045-038 97045-718 97045-714 97045-838 97044-976 97044-972 97045-284 97045-166 97045-280 89231-882 30618-224 97045-842 97045-204 97045-722 30618-228 89231-886 30618-346 97045-602 97045-200 89231-802 97045-846 97045-726 97045-606 30618-260 97044-960 30618-252 97045-296 30618-374 97045-054 89231-870 97045-170 30618-256 30618-378 97045-050 97045-216 89231-878 97045-734 97045-614 30618-418 97045-850 97045-858 97045-738 97045-618 97044-956 30618-370 97045-220 89231-862 97045-622 97045-502 97045-228 97045-866 30618-248 30618-402 89231-866 97045-586 97045-862 97045-466 97045-224 30618-406 97045-742 97045-626 97045-506 97045-070 30618-280 97045-110 97045-594 30618-272 30618-394 97045-474 89231-850 30618-276 30618-430 30618-398 97045-074 97045-634 97045-118 97045-514 97045-750 89231-858 97045-114 97045-598 97045-478 97045-758 97045-638 97045-518 97045-240 97045-482 97045-086 30618-382 97045-000 97045-122 30618-300 30618-422 89231-962 97045-248 97045-402 30618-304 97045-766 89231-842 30618-426 97045-646 97045-008 89231-966 97045-526 97045-486 30618-308 97045-244 89231-846 97045-762 97045-642 97045-004 97045-522 97045-406 97045-090 30618-296 97045-098 30618-330 97045-494 97045-252 97045-770 30618-212 97045-094 30618-334 89231-794 89231-830 89231-950 97045-534 89231-798 97045-414 97045-778 97045-658 97045-498 97045-136 89231-958 97045-654 89231-818 89231-938 97045-538 97045-418 97044-996 97045-020 89231-780 97045-782 30618-284 97045-386 89231-784 30618-320 30618-442 97045-140 89231-942 30618-324 89231-822 30618-446 97045-024 97045-266 89231-946 97045-662 97045-542 97045-300 97045-422 30618-208 89231-826 97045-786
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and Caps, Assembled, DWK Life Sciences
Microplates Deep Well Microplates
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with glass inserts and color-coded caps.
  • Variety of volumes available
  • Standard chromatography closures
  • Caps with septa
  • Silicone mats are resealable
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary, reseal after the sample has been injected. Users may also choose the security of a stopper coupled with the strength of a mechanical closure system. The mechanical closure system allows even GC samples to be converted to 96-Well format, by sealing the limited volume insert on the inside surface of the insert and compressing the PTFE/Silicone rubber using a mechanical seal. A variety of insert options allows users to choose the specifications appropriate for any application.
The system combines microplates, glass inserts, μLmats™, and caps with chromatography-based septa to offer secure vessels to maintain even the most volatile samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty