Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

WHEATON® MicroLiter 9 mm Screw-Thread Vials, DWK Life Sciences

Supplier: DWK Life Sciences
09-1300-100 09-1400-S 09-1200 09-1200-101 09-1000-140 09-1000-100 09-1200-102 76-1200-S 76-2200-300 76-1200 09-1200-100 09-3200 09-2200 09-2200-102 09-1300-100B 09-2200-300 76-1200-100 09-1000-101 09-1000-300 09-1300-1000 76-2200-S 09-2200-101 09-1000 76-2200 09-1000-S 76-1200-101 09-1200-S 09-2200-S 76-1200-300 09-1200-300 09-2300-1000 76-2200-101 09-1400
97046-494PK 44.26 USD InStock
97046-494 97046-498 97046-312 97046-752 97046-496 10039-382 97046-750 97046-310 97046-678 97046-316 97046-676 97048-414 97046-314 97048-415 97046-748 97046-746 97046-926 97046-682 89496-144 89496-146 97046-680 97046-320 97046-502 97046-500 97048-228 97048-236 97048-412 97048-410 97048-234 97046-934 97046-318 97048-232 97048-230
WHEATON® MicroLiter 9 mm Screw-Thread Vials, DWK Life Sciences
Vials
All vials may appear to be created equal, but quality and performance make the difference when analyzing vital samples. MicroLiter vials are identical, clean, and dependable, and are certified to ensure optimal performance and reproducible results.

  • Performance equal to an 11 mm crimp-top vial
  • Inserts available

MicroLiter vials are cleaned and packaged in a certified Class 10000 clean room according to a designed process, developed to eliminate chromatographic errors associated with vials

The vials and inserts are made from Type 1 borosilicate glass or polypropylene, and meet strict tolerances. Their performance is equal to an 11 mm crimp-top vial, with the convenience and accessibility of screw threads. The µl Vials™ feature conical bottoms. The 76-Series Vials™ feature a modified neck finish to ensure a consistent, repeatable seating of the gripper against the bottom of the cap.

Critical physical dimensions are fully tested, as well as chemical cleanliness testing to ensure that trace levels of residues are not present.

Certifications: Certified Clean

Order Now
 

ORDER

COMPLEMENTARY PRODUCTS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty