Would you like to log in again?
Limited Volume Vial Inserts, MicroLiter
Supplier: DWK Life Sciences
11-0000-100-S 11-0000-300 11-0000-300-B 11-0000-101-B 11-0000-100-B 11-0000-101 11-0000-300-S 11-0000-100 11-0000-101-S 11-0000-140
97047-008PK 225.14 USD InStock
97047-008 97047-020 97047-010 97047-022 97047-012 97047-024 97047-014 97047-026 97047-016 97047-018
Limited Volume Vial Inserts, MicroLiter
Vials
Inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances.
  • Conical and flat bottom
  • Meet strict tolerances
Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty