Would you like to log in again?
WHEATON® MicroLiter Crimp-Top Headspace Vials, 20 mm, DWK Life Sciences
Supplier: DWK Life Sciences
20-1400 20-1100 20-2000-S 20-2100-S 20-2400 20-1000-S 20-1000 20-2100 20-2000 20-6000 20-5000 20-2300
89498-624PK 79.39 USD InStock
89498-624 97047-998 97047-988 97047-978 97047-990 97047-980 97047-992 97047-982 97047-994 97047-984 97047-996 97047-986
WHEATON® MicroLiter Crimp-Top Headspace Vials, 20 mm, DWK Life Sciences
Vials
These headspace vials offer excellent performance.
Caps are blue polypropylene. Butyl stoppers can be used when PTFE is not required. Butyl/PTFE septa are an economical choice with low background peaks. Molded PTFE/butyl septa offer the tightest seal of all PTFE items, as well as low background peaks. PTFE/silicone septa are excellent for volatiles. Aluminum/silicone septa are also excellent for volatiles, but not suitable for repeat injections. Stoppers and septa with butyl are usable from –40 to 120 °C, PTFE/silicone from –60 to 200 ºC, and aluminum/silicone from –60 to 120 °C. AlumiTin™ seals combine the ease of use of aluminum with the magnetic characteristics of tin.
Every glass vial is precleaned and packaged in an ISO Class 7 (Class 10000) cleanroom, in a box bearing the Class 10K seal. The result is a vial that exceeds most OEM requirements at no additional cost. Cleaner vials improve performance by eliminating almost all particulates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty