Would you like to log in again?
Portable Turbidimeter, Model 2100Q, Hach
Supplier: Hach
2100Q01 2100Q01
97049-281EA 4372.39 USD InStock
97049-281 CA97049-280
Portable Turbidimeter, Model 2100Q, Hach
Turbidity Meters
The 2100Q Portable Turbidimeter offers unsurpassed ease of use and accuracy in turbidity measurement.
  • Portable
  • Easy to use
  • Advanced features
  • One year manufacturer's warranty
Only Hach offers this unique combination of advanced features and measurement innovation, giving you accurate results every time
Easy on-screen assisted calibration and verification. Simple data transfer. Accurate for rapidly settling samples. Convenient data logging. Optical system for precision in the field. Tungsten filament lamp light source. Selectable on/off signal averaging.
Measurement method is ratio turbidimetric determination using a primary nephelometric light scatter signal (90°) and transmitted light scatter signal. Measurement modes include Normal (Push to Read), Signal Averaging, and Rapidly Settling Turbidity. User interface languages include English, French, German, Italian, Spanish, Portuguese (BR), Portuguese (PT), Bulgarian, Chinese, Czech, Danish, Dutch, Finnish, Greek, Hungarian, Japanese, Korean, Polish, Romanian, Russian, Slovenian, Swedish, and Turkish.
Compliant with USEPA Method 180.1 design criteria.
Storage conditions: -40 to 60 °C
Certifications: CE/RoHS/WEEE. IP67 Enclosure Rating.
Ordering information: One-year warranty. Optional USB interface. Includes instrument, case assembly, 4 AA alkaline batteries, 6 sample cells, StablCal ampule kit, silicone oil, oiling cloth, and manual.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty