Designed for maximum product recovery for lab-scale or scale-up process volumes up to 4 L.
- Membrane Cassette Filter Media: Omega™ (low protein-binding, modified PES) membranes
- All product-wetted parts comply with pharmaceutical standards
- Easy connections through luer lock fitted ports with polished 316L stainless steel to ensure the same compatibility characteristics as production-scale Centrasette holders
Scale-up/scale-down device for development of tangential flow filtration (TFF) processes, biopharmaceutical small batch production, scale-down investigations, and production of materials for safety and efficacy studies (Phase I clinicals).
Identical path lengths provide linear scale-up to Pall Centramate and Centrasette™ cassette holders. The LV Centramate holder may be attached to other laboratory TFF systems to improve their scalability characteristics.
Low hold-up volume allows high concentration factors to be achieved from small starting volumes. A complete TFF system scalable to full production plant can be optimized using the volumes typically generated in the development or discovery lab.