Would you like to log in again?
LV Centramate™ Lab Tangential Flow Filtration System, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Centramate™
OS010T02 OS001T12 OS300T02 OS030C11P2 OS030T12 OS001T12 OS300C11P2 CM018LV OS005T12 OS030C11P2 OS050T02 OS070T12 OS030T12 CM018LV OS300T12 OS001T02 OS070T02 OS005C11P2 OS050C11P2 OS030T02 OS005C11P2 OS300C11P2 OS005T02 OS100T12 OS050T02 OS005T12 OS100T12 OS001C11 OS010T02 OS010T12 OS003T12 OS050T12 OS300T12 OS001T02 OS010T12 OS300T02 OS030T02 OS005T02 OS100T02 OS100T02 OS050C11P2 OS050T12 OS001C11
97049-464EA 1143.56 USD InStock
CA97049-474 97049-464 CA97049-454 97049-466 CA97049-476 97049-460 CA97049-470 97049-482 97049-484 CA97049-472 97049-462 97049-480 87002-092 76417-794 CA87002-080 CA97049-466 29301-432 97049-458 CA87002-104 CA97049-468 97049-454 97049-476 CA97049-462 CA97049-484 CA29299-018 97049-455 97049-456 97049-478 97049-472 CA97049-460 97049-474 CA97049-482 97049-470 87002-080 CA76417-794 87002-088 CA29900-082 87002-104 CA29301-432 CA97049-478 97049-468 CA97049-456 CA97049-458
LV Centramate™ Lab Tangential Flow Filtration System, Cytiva (Formerly Pall Lab)
Filters Tangential Flow Cassettes
Designed for maximum product recovery for lab-scale or scale-up process volumes up to 4 L.
  • Membrane Cassette Filter Media: Omega™ (low protein-binding, modified PES) membranes
  • All product-wetted parts comply with pharmaceutical standards
  • Easy connections through luer lock fitted ports with polished 316L stainless steel to ensure the same compatibility characteristics as production-scale Centrasette holders
Scale-up/scale-down device for development of tangential flow filtration (TFF) processes, biopharmaceutical small batch production, scale-down investigations, and production of materials for safety and efficacy studies (Phase I clinicals).
Identical path lengths provide linear scale-up to Pall Centramate and Centrasette™ cassette holders. The LV Centramate holder may be attached to other laboratory TFF systems to improve their scalability characteristics.
Low hold-up volume allows high concentration factors to be achieved from small starting volumes. A complete TFF system scalable to full production plant can be optimized using the volumes typically generated in the development or discovery lab.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty