Would you like to log in again?
Supracap™ 100 Depth Filter Capsules, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
NP6P1001 NP6PDH46 NP5LP2506 NP6PDK56 NP6B0201 NP5LPDD11 NP6P2506 NP5LP9001 NP6PDK56 NP5LB0206 NP5LPDK51 NP6PDD11 NP6PDD16 NP5LP7001 NP5LPDE21 NP5LP2001 NP6P1006 NP5LP2006 NP6P2501 NP6P2001 NP5LPDK56 NP5LP1006 NP5LP7001 NP6P9001 NP6PDE26 NP6PDH41 NP6P1001 NP5LP2506 NP5LPDK56 NP6P2006 NP5LPDD16 NP5LPDP87 NP6P2506 NP6P7001 NP6PDH46 NP6PDK51 NP5LPDH41 NP6P7006 NP6B0201 NP6PDP86 NP6PDD16 NP5LB0201 NP5LP9006 NP6P7006 NP6P1006 NP6PDE26 NP5LP1001 NP6PDK51 NP5LP1006 NP6PDP81 NP5LPDK51 NP5LP9001 NP5LPDE26 NP5LP2006 NP6PDE21 NP6P9001 NP5LPDE21 NP5LPDH46 NP5LPDH41 NP6P9006 NP5LB0201 NP6PDE21 NP5LPDD16 NP5LP2501 NP6P2501 NP6PDH41 NP6P7001 NP5LP2501 NP5LP7006 NP5LPDE26 NP5LP2001 NP5LPDH46 NP6P2001 NP5LP9006 NP5LP1001 NP6PDD11 NP6P9006 NP6P2006 NP5LP7006 NP5LPDD11
89167-280EA 522.19 USD InStock
89167-280 CA97050-022 CA97050-026 CA97050-024 97048-716 CA97050-028 97048-714 97048-712 CA97048-694 CA89167-258 CA89167-256 CA89167-254 CA89167-252 89167-282 89167-270 97050-044 97048-708 CA97048-716 97048-706 76285-220 97048-704 CA97048-714 CA97048-712 97048-702 CA97048-710 97048-700 76285-222 97048-710 89167-278 89167-276 97050-042 CA89167-282 89167-274 97050-040 CA89167-280 89167-272 CA97050-044 97050-036 CA97050-042 97050-034 CA97048-708 76285-218 97050-038 CA97048-706 CA97048-704 CA97048-702 CA97048-700 89167-268 CA89167-278 CA89167-276 89167-266 89167-264 CA89167-274 CA89167-272 89167-262 CA97050-040 97050-032 CA89167-270 89167-260 97050-030 CA97050-034 CA97050-032 97050-022 97050-028 CA97050-038 97050-026 CA97050-036 CA97048-698 97048-698 97048-694 CA89167-268 89167-258 CA89167-266 CA89167-264 89167-256 CA89167-262 89167-254 CA97050-030 89167-252 CA89167-260
Supracap™ 100 Depth Filter Capsules, Cytiva (Formerly Pall Lab)
Filters Capsule Filters
Supracap™ 100 depth filter capsules are ideal prefilters made with Seitz® high performance depth filter media for the removal of unwanted solids from solutions in laboratory and process development applications.
  • Scalable
  • Low hold-up volume
  • Increased recovery
  • Three grades available
Supracap 100 capsules feature low hold-up volume that allows for increased product recovery and that requires low post-use rinse volumes.
Supracap 100 capsules are offered in three depth filter grades: Seitz Bio 20, Seitz P-series, and Seitz HP-series.
The Seitz Bio 20 media grade is made of highly purified natural and modified celluloses and is free from inorganic materials. Seitz Bio 20 media is ideal for ion sensitive products such as therapeutic proteins, dialysis solutions, and protein-based diagnostics.
Seitz P-series media are comprised of cellulose fibers, filter aids (diatomaceous earth and perlite), and resins. This grade is recommended for coarse filtration in the biotechnological and pharmaceutical industries.
The typical sample volume for the Supracap 100 depth filter capsules is 3 to 100 liters.
Certifications: All plastic components used in construction meet the specifications for Biological Reactivity Tests In Vivo for Class VI Plastics (121 °C) as described in the current United States Pharmacopoeia (USP).
Ordering information: Supracap 100 capsules are available in 5 and 10" sizes. Housing head and bowl are made of polypropylene. O-rings are made of silicon elastomers.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty