Would you like to log in again?
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11-0054N 11-0055NT 11-0055K 11-0051KT 11-0053DBT 11-0053N-B 11-0053BT 11-0054K-B 11-0056N 11-0055DB 11-0053R-B 11-0051G-B 11-0054BT 11-0055R 11-0056DB 11-0054N-B 11-0055BT 242776-05 11-0051R 11-0055N-B 11-0055G 11-0053K 242772-05 11-0051B-B 11-0052DB-B 11-0056R-B 11-0052K 11-0052K-B 11-0054KT 11-0054DB 11-0053R 11-0055DB-B 11-0052B-B 11-0052DB 11-0054K 11-0053N 11-0054RT 11-0052N 11-0052N-B 11-0054B-B 11-0054NT 11-0051NT 242776-04 11-0054G 11-0052DBT 11-0054R-B 11-0051R-B 11-0053DB-B 11-0052G 11-0055GT 11-0051BT 11-0056N-B 11-0053K-B 242772-04 11-0055K-B 11-0055B-B 11-0055DBT 11-0053B-B 242772-06 11-0051N-B 11-0053NT 11-0051N 11-0053G-B 11-0056DB-B 242776-02 11-0053GT 11-0054DBT 11-0054GT 11-0053G 11-0055KT 11-0051K 11-0052NT 11-0052RT 11-0055G-B 11-0051DB 11-0051B 11-0056G 11-0054DB-B 11-0053B 11-0056G-B 11-0052R 11-0055B 11-0051G 11-0051RT 11-0051K-B 11-0053KT 11-0053DB 11-0052B 11-0056R 11-0055RT 11-0052GT 11-0052R-B 242776-01 11-0054B 11-0055R-B 11-0056B 11-0054R 11-0056K 11-0053RT 11-0056K-B 11-0051GT 11-0052KT 11-0055N 11-0051DBT 11-0051DB-B 242772-01 11-0054G-B 11-0052G-B 242772-02
97047-160PK 56.73 USD InStock
97047-160 97047-162 97047-164 97047-166 97047-200 97047-168 97047-202 97047-204 89235-558 97047-150 97047-152 97047-154 97047-156 97047-158 97050-398 97050-399 97047-218 97050-396 89235-564 97050-397 89235-568 89235-562 89235-560 97047-180 10027-474 97047-182 10027-472 97047-184 10027-476 97047-186 97047-220 97050-390 97047-188 97050-391 97047-222 97050-394 97047-224 97050-395 97050-392 97047-226 97050-393 97050-387 97047-206 97050-388 89235-574 97047-208 97050-385 97050-386 97050-389 89235-572 89235-570 66014-630 10027-464 97047-170 97047-172 97047-174 10027-466 97047-176 97047-210 97047-178 97047-212 97050-383 97047-214 97050-384 97050-382 97047-216 97047-118 97050-410 97050-413 97050-414 97050-411 97050-412 97050-417 97050-415 97050-416 97047-120 97047-122 97047-124 97047-126 97047-128 97047-228 97050-402 97050-403 97050-400 97050-401 97050-406 97050-407 97050-404 97050-405 97050-408 97050-409 97047-190 97047-192 97047-194 97047-196 97047-230 97047-198 97047-232 97047-114 97047-116 97047-142 97047-144 97047-146 97047-148 97047-130 97047-132 97047-134 97047-136 97047-138
WHEATON® MicroLiter 11 mm Snap-Top Caps, DWK Life Sciences
Closures Chromatography Vial Closures
These snap-top caps eliminate the need for crimping and decapping tools.
  • Manufactured from PE
  • Fits the WHEATON E-Z Vials® with snap ring top
Available in multiple colors and septa types.
Order Now
ORDER
COMPLEMENTARY PRODUCTS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty