Would you like to log in again?
Glass Solvent Bottle with ITSP Connector, 1 L, MicroLiter, DWK Life Sciences
Supplier: DWK Life Sciences
LPAL.1000ML
97050-590EA 213.4 USD InStock
97050-590
Glass Solvent Bottle with ITSP Connector, 1 L, MicroLiter, DWK Life Sciences
Bottles Solution Bottles
Glass solvent bottle is 1 L and has an 8 mm connector for HTS, HTS-xt, HTC, HTC-xt, HTX, and HTX-xt PAL systems.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty