Would you like to log in again?
MICROBEE C18 4.6 X 150MM STD
Supplier: MicroSolv Technology Corp
Hplc Column^ Cogent Microbee C18 10Um Irregular Shaped 125A 4.6Mm X 150Mm Standard End Fittings. Compare To Waters Microbondak.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MICROBEE C18 4.6 X 150MM STD 97050-986 59018-15P Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty