Would you like to log in again?
Cogent HPS™ C8 HPLC Column, MicroSolv
Supplier: MicroSolv
75008-05P-1 75008-05P-3 75008-15P-1 75008-10P-1 75008-10P-2 75008-05P-2 75008-10P 75008-15P-2 75008-25P 75008-7.5P 75008-15P 75008-25P-1
97051-286EA 935.61 USD InStock
97051-286 97051-296 97051-078 97051-288 97051-298 97051-300 97051-080 97051-082 97051-290 97051-292 97051-084 97051-294
Cogent HPS™ C8 HPLC Column, MicroSolv
Chromatography Columns
Cogent HPS™ columns are double end-capped and have narrow particle distribution making them extremely high-quality, Type-B, silica-based HPLC columns.
Used for routine applications worldwide, these columns offer great efficiency, selectivity and value. Bonding is monofunctional which provides greater stability and reproducibility.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty