Would you like to log in again?
VIALS CLEAR WITH WRITE ON
Supplier: MicroSolv Technology Corp
Vials^ Autosampler 1.5Ml 12X32Mm Serum Finish/ Crimp Top With Wide Cpening Clear Borosilicate Glass With Write On Patch. Basik Brand. 100/Pack.Microsolv Cat. No.9502C-0-Wcv
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VIALS CLEAR WITH WRITE ON 97052-314 9502C-0-WCV Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty