Would you like to log in again?
AQ™ Advanced Quality Borosilicate Glass and Plastic Inserts
Supplier: MicroSolv
RSA-Pro™ AQ™ RSA™ BASIK™ MicroSolv™
9504S-03ND 9502S-02ND 9503S-05N 9502S-03ND 9502S-02PP-K 9502S-02L-K 9502S-02N-RS 9504S-01ND 95001-01N 95001-INS 9502S-02CP 9502C-0-FB 9504S-02ND 9502S-05N-K 9504S-02N 9502S-04N 9504S-03N 9502S-05ND 9502S-02E 9502S-05ND-K 9502S-05N 9502S-05N-A 9502S-02ND-RS 95001-03N 9504S-04N 95001-02N 9502S-02CP-K 9502S-03E 95001-01ND 95001-04ND 9502S-04ND 9502C-0-FB-K 9504S-04ND 9502S-02N 9502S-02PP 9504S-01N 95001-02ND 9503S-05FB 9502S-03N 9504C-INS 95001-04N 95001-03ND
97055-356PK 119.11 USD InStock
97055-356 97055-358 10804-674 97055-350 97055-352 97055-354 97054-534 97052-307 97054-532 97052-768 97051-400 97052-448 97054-538 97051-402 97052-304 97052-766 97052-446 97054-558 97054-536 97052-444 97052-762 97052-442 97052-760 97051-408 97054-552 97052-440 97054-530 97055-344 97055-346 97055-348 10060-156 97055-340 97051-396 97051-398 97052-438 97054-528 97051-410 97054-526 97052-458 97052-456 97054-540 97052-450
AQ™ Advanced Quality Borosilicate Glass and Plastic Inserts
Vials
Low volume glass and plastic precision manufactured inserts for HPLC, GC, LCMS and GCMS.
These low volume inserts for autosampler vials are best in class because of the quality of the starting glass and plastics used to produce precision shapes, for high performance results. Inject very small volumes from the inserts to maximize the number of injections per insert/vial. Choose from economical options including flat bottom or conical bottom. Precision point self aligning inserts or inserts with attached plastic springs are available.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty