Would you like to log in again?
CAPS SCREW TOP PRESLIT SEPTA
Supplier: MicroSolv Technology Corp
Caps^ Autosampler, Screw Top, White With Bonded Preslit Silicone/Ptfe Septa. For Use With 9-425Mm, 12X32Mm Vials. Microsolv Brand. 100/Pkmicrosolv Cat. No.9502S-30C-W
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS SCREW TOP PRESLIT SEPTA 97052-536 9502S-30C-W Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty