Would you like to log in again?
EPP MAX REC VIALS AMBER & CAPS
Supplier: MicroSolv Technology Corp
Vials^ Max Recovery, Screw Top With 9-425Mm Threads, 1.5Ml Fill Volume 12X32Mm, Deactivated. Clear Borosilicate Glass. Caps Sold Separately. 100/Pack.Microsolv Cat. No.9512S-0Dcv
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EPP MAX REC VIALS AMBER & CAPS 97052-608 9512S-1AP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty