Would you like to log in again?
EPP MAXREC VIAL CLR SILAN CAPS
Supplier: MicroSolv Technology Corp
Easy Purchase Pack^ Mrq Maximum Recovery Screw Top Autosampler Vials Clear 1.2Ml Fill Volume, 12X32Mm And Blue Screw Caps With Preslit Bonded Silicone/Ptfe Septa. 100/Pack.Microsolv Cat. No.9512S-3Cp-T
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EPP MAXREC VIAL CLR SILAN CAPS 97052-624 9512S-3DCP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty