Would you like to log in again?
Fused-Insert Vials, Screw Caps w/ Pre-Slit Septa, MicroSolv™
Supplier: MicroSolv
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9532S-0CV 9532S-3CP-WT 9532S-3DCP 9532S-3CP
97057-792PK 305.62 USD InStock
97057-792 97052-668 89220-070 97057-798
Fused-Insert Vials, Screw Caps w/ Pre-Slit Septa, MicroSolv™
Vials
These vials have a small volume insert installed and permanently 'fused' in.
These vials/inserts are available for snap/crimp top, 9 mm screw top vials. The ultimate in convenience when using small volumes. These vials have screw caps with PTFE/Silicone septa.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty