Would you like to log in again?
CAPS SNAP RED WHITE PTFE
Supplier: MicroSolv Technology Corp
Caps^ Autosampler, Snap Top, Red With White Ptfe Septa. For Use With 12X32Mm Snap Top Vials. Microsolv Brand. 1000/Case.Microsolv Cat. No.9502C-50Cr-M
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
CAPS SNAP RED WHITE PTFE 97052-740 9502C-50CR-M Case of 1,000 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty